missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabutylammonium fluoride, 75% solution in water, Thermo Scientific Chemicals
68.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Density | 0.9530g/mL |
Packaging | Glass Bottle |
Linear Formula | [CH3(CH2)3]4NF |
Quantity | 100 g |
Merck Index | 15,9332 |
Specific Gravity | 0.953 |
IUPAC Name | tetrabutylazanium fluoride |
Formula Weight | 261.46 |
Chemical Name or Material | Tetrabutylammonium Fluoride |
Safety and Handling
Product Identifier
- Tetrabutylammonium fluoride, 75% solution in water
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H301-Toxic if swallowed.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'