missing translation for 'onlineSavingsMsg'
Learn More

Tetrabenzyl pyrophosphate, 98%, Thermo Scientific Chemicals

Catalog Number p-7045007
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
990-91-0
C28H28O7P2
538.473
MFCD00051941
NSBNXCZCLRBQTA-UHFFFAOYSA-N
tetrabenzyl pyrophosphate, tetrabenzyl diphosphate, pyrophosphoric acid tetrabenzyl ester, tetrabenzylpyrophosphate, diphosphoric acid, tetrakis phenylmethyl ester, dibenzyl dibenzyloxyphosphoryl oxyphosphonate, benzyl pyrophosphate, acmc-209sbp, tetra benzyl pyrophosphate, tetrabenzyl diphosphate #
563183
dibenzyl bis(phenylmethoxy)phosphoryl phosphate
C1=CC=C(C=C1)COP(=O)(OCC2=CC=CC=C2)OP(=O)(OCC3=CC=CC=C3)OCC4=CC=CC=C4
This item is not returnable. View return policy
990-91-0
C28H28O7P2
538.473
MFCD00051941
NSBNXCZCLRBQTA-UHFFFAOYSA-N
tetrabenzyl pyrophosphate, tetrabenzyl diphosphate, pyrophosphoric acid tetrabenzyl ester, tetrabenzylpyrophosphate, diphosphoric acid, tetrakis phenylmethyl ester, dibenzyl dibenzyloxyphosphoryl oxyphosphonate, benzyl pyrophosphate, acmc-209sbp, tetra benzyl pyrophosphate, tetrabenzyl diphosphate #
563183
dibenzyl bis(phenylmethoxy)phosphoryl phosphate
C1=CC=C(C=C1)COP(=O)(OCC2=CC=CC=C2)OP(=O)(OCC3=CC=CC=C3)OCC4=CC=CC=C4

Tetrabenzyl pyrophosphate is used in the preparation of phosphoryl derivatives of shikimic acid in the presence of LDA. It is used in the preparation of dibenzyl phosphoro fluoridate in the presence of cesium fluoride as a catalyst. It is also used as a precursor in pharmaceuticals and involved in the phosphorylation of inositol derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrabenzyl pyrophosphate is used in the preparation of phosphoryl derivatives of shikimic acid in the presence of LDA. It is used in the preparation of dibenzyl phosphoro fluoridate in the presence of cesium fluoride as a catalyst. It is also used as a precursor in pharmaceuticals and involved in the phosphorylation of inositol derivatives.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acid and bases and reducing agents.
TRUSTED_SUSTAINABILITY
Melting Point 58°C to 64°C
Quantity 250 mg
UN Number UN3261
Beilstein 2068292
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 538.48
Percent Purity 98%
Chemical Name or Material Tetrabenzyl pyrophosphate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'