missing translation for 'onlineSavingsMsg'
Learn More

Tetraammineplatinum(II) hydrogen phosphate solution, Pt 2% w/w (cont. Pt), Thermo Scientific™

Catalog Number p-7059090
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
127733-98-6
H13N4O4PPt
359.19
MFCD03788258
LSDWMQXWIQMHNQ-UHFFFAOYSA-L
azane;hydrogen phosphate;platinum 2+, platinum 2+ tetraamine hydrogen phosphate, platinum 2+ hydrogen phosphate-ammonia 1/1/4, platinum 2+ ion tetraamine hydrogen phosphate
platinum(2+) tetraamine hydrogen phosphate
N.N.N.N.[Pt++].OP([O-])([O-])=O
This item is not returnable. View return policy
127733-98-6
H13N4O4PPt
359.19
MFCD03788258
LSDWMQXWIQMHNQ-UHFFFAOYSA-L
azane;hydrogen phosphate;platinum 2+, platinum 2+ tetraamine hydrogen phosphate, platinum 2+ hydrogen phosphate-ammonia 1/1/4, platinum 2+ ion tetraamine hydrogen phosphate
platinum(2+) tetraamine hydrogen phosphate
N.N.N.N.[Pt++].OP([O-])([O-])=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Colorless
Quantity 1 g
Solubility Information Fully miscible with water.
Formula Weight 359.18 (Anhydrous)
Concentration or Composition (by Analyte or Components) Pt 2% w/w (cont. Pt)
Physical Form Liquid
Chemical Name or Material Tetraammineplatinum (II) hydrogen phosphate solution
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'