missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Tetraammineplatinum(II) chloride solution, Pt 9-11% w/w (cont. Pt)

Brand:  Alfa Aesar™ 044045.NB

Code : NEW

Additional Details : CAS Number : 13933-32-9 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11440660

  • £620.31 / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Tetraammineplatinum (II) chloride solution
H12Cl2N4Pt
334.11 (Anhydrous)
13933-32-9
[Pt(NH3)4]Cl2
MFCD00011475

Specifications

Tetraammineplatinum (II) chloride solution
Liquid
[Pt(NH3)4]Cl2
MFCD00011475
Pt 9 to 11% w/w (cont. Pt)
13933-32-9
H12Cl2N4Pt
334.11 (Anhydrous)
Colorless to pale yellow
5g
Videos
SDS