missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Tetraamminepalladium(II) tetrachloropalladate(II)

Brand:  Alfa Aesar™ 039447.03

Code : NEW

Additional Details : CAS Number : 13820-44-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 11348238

  • POA / 1g

This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.


Specifications

Chemical Identifiers

Tetraamminepalladium(II) tetrachloropalladate(II)
13820-44-5
422.8
MFCD00049783

Specifications

Tetraamminepalladium(II) tetrachloropalladate(II)
2.489g/mL
422.8
192°C (dec.)
1g
13820-44-5
Powder
MFCD00049783
Odorless
Videos
SDS