Learn More
Alfa Aesar™ Tetraamminepalladium(II) hydrogen carbonate
Brand: Alfa Aesar™ 045082.14
Code : NEW
Additional Details : CAS Number : 134620-00-1 Weight : 0.02500kg Transport : UN number : 3077 Chem class : 1 Pack group : III
Quantity | 25g |
---|
Chemical Identifiers
Tetraamminepalladium (II) hydrogen carbonate | |
C2H14N4O6Pd | |
296.58 | |
UN3077 |
134620-00-1 | |
[Pd(NH3)4](HCO3)2 | |
MFCD09953449 |
Specifications
Tetraamminepalladium (II) hydrogen carbonate | |
2.04g/mL | |
C2H14N4O6Pd | |
296.58 | |
UN3077 | |
25g |
134620-00-1 | |
Crystalline | |
[Pd(NH3)4](HCO3)2 | |
MFCD09953449 | |
Colorless to light yellow |