missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Tetraamminedioxoosmium(VI) chloride, Os 52.2% min

Brand:  Alfa Aesar™ 041020.03

Code : NEW

Additional Details : CAS Number : 18496-70-3 Weight : 0.00100kg

Product Code. 15401697

  • £253.76 / 1g


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Specifications

Tetraamminedioxoosmium(VI) chloride
Powder
MFCD00798548
1g
18496-70-3
361.23
Os 52.2% min.
Videos
SDS