missing translation for 'onlineSavingsMsg'
Learn More

Tetraallylsilane, 97%, Thermo Scientific Chemicals

Catalog Number p-7058755
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1112-66-9
C12H20Si
192.377
MFCD00048161
AKRQMTFHUVDMIL-UHFFFAOYSA-N
tetraallylsilane, silane, tetra-2-propenyl, tetrakis prop-2-en-1-yl silane, tetrakis prop-2-enyl silane, silane, tetraallyl, tetraallyl silane, zlchem 610, acmc-20ajk5, tetra 2-propenyl silane, silane,tetra-2-propen-1-yl
565641
tetrakis(prop-2-enyl)silane
C=CC[Si](CC=C)(CC=C)CC=C
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1112-66-9
C12H20Si
192.377
MFCD00048161
AKRQMTFHUVDMIL-UHFFFAOYSA-N
tetraallylsilane, silane, tetra-2-propenyl, tetrakis prop-2-en-1-yl silane, tetrakis prop-2-enyl silane, silane, tetraallyl, tetraallyl silane, zlchem 610, acmc-20ajk5, tetra 2-propenyl silane, silane,tetra-2-propen-1-yl
565641
tetrakis(prop-2-enyl)silane
C=CC[Si](CC=C)(CC=C)CC=C

Synthesis of dendrimers

It is used in synthesis of dendrimers and as an intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 91°C (10mmHg)
Physical Form Liquid
Quantity 1 g
Flash Point 77°C (171°F)
Solubility Information Insoluble in water.
Formula Weight 192.38
Percent Purity 97%
Refractive Index 1.485
Density 0.8314 g/mL
Chemical Name or Material Tetraallylsilane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'