missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetra-n-propyltin, Thermo Scientific Chemicals
121.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
Description
It can be used to produce Dichlor-dipropyl-stannan and propane-1-sulfinyl chloride at room temperature. Applications include preservation of wood, textiles, leather, and paper.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS | 2176-98-9 |
Color | Colorless |
Density | 1.11 g/mL |
Boiling Point | 222°C |
Odor | Unpleasant |
Molecular Formula | C12H28Sn |
Linear Formula | (CH3CH2CH2)4Sn |
Refractive Index | 1.4745 |
MDL Number | MFCD00015196 |
Quantity | 10 g |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'