missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-propylammonium bromide, 98%, Thermo Scientific Chemicals

Catalog Number p-7081516
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
1941-30-6
C12H28BrN
266.267
MFCD00011840
BGQMOFGZRJUORO-UHFFFAOYSA-M
tetrapropylammonium bromide, tetra-n-propylammonium bromide, 1-propanaminium, n,n,n-tripropyl-, bromide, tetrapropylazanium bromide, tpabr, n,n,n-tripropyl-1-propanaminium bromide, n,n,n-tripropylpropan-1-aminium bromide, tetrapropylammonium bromide tpabr, 1-propanaminium, n,n,n-tripropyl-, bromide 1:1, tetra propyl ammonium bromide
74745
CHEBI:55318
tetrapropylazanium;bromide
CCC[N+](CCC)(CCC)CCC.[Br-]
This item is not returnable. View return policy
1941-30-6
C12H28BrN
266.267
MFCD00011840
BGQMOFGZRJUORO-UHFFFAOYSA-M
tetrapropylammonium bromide, tetra-n-propylammonium bromide, 1-propanaminium, n,n,n-tripropyl-, bromide, tetrapropylazanium bromide, tpabr, n,n,n-tripropyl-1-propanaminium bromide, n,n,n-tripropylpropan-1-aminium bromide, tetrapropylammonium bromide tpabr, 1-propanaminium, n,n,n-tripropyl-, bromide 1:1, tetra propyl ammonium bromide
74745
CHEBI:55318
tetrapropylazanium;bromide
CCC[N+](CCC)(CCC)CCC.[Br-]

Tetra-n-propylammonium bromide, is used as surface-active agents, Solvents, Intermediates, Active Ingredient for Conditioners, Antistatic Agent, Detergent Sanitisers, Softner for textiles and paper products, Phase Transfer Catalyst, Antimicrobials, Disinfection Agents And Sanitizers, Slimicidal Agents, Emulsifying Agents and Pigment Dispersers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetra-n-propylammonium bromide, is used as surface-active agents, Solvents, Intermediates, Active Ingredient for Conditioners, Antistatic Agent, Detergent Sanitisers, Softner for textiles and paper products, Phase Transfer Catalyst, Antimicrobials, Disinfection Agents And Sanitizers, Slimicidal Agents, Emulsifying Agents and Pigment Dispersers.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatabile to Strong oxidizing agents. Stable under ordinary conditions (Hygroscopic)
TRUSTED_SUSTAINABILITY
Melting Point ∼270°C (decomposition)
Quantity 100 g
Beilstein 3567846
Sensitivity Hygroscopic
Solubility Information It is soluble in water.
Formula Weight 266.27
Percent Purity ≥98%
Chemical Name or Material Tetra-n-propylammonium bromide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'