missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-octylammonium bromide, 98+%, Thermo Scientific Chemicals

Catalog Number p-7022246
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
14866-33-2
C32H68BrN
546.807
MFCD00011863
QBVXKDJEZKEASM-UHFFFAOYSA-M
tetraoctylammonium bromide, tetra-n-octylammonium bromide, tetraoctylazanium bromide, tetraoctyl ammonium bromide, 1-octanaminium, n,n,n-trioctyl-, bromide, unii-52qw29owoc, 52qw29owoc, tetra n-octyl ammonium bromide, acmc-1bvth, tetra-octylammonium bromide
2734117
tetraoctylazanium;bromide
CCCCCCCC[N+](CCCCCCCC)(CCCCCCCC)CCCCCCCC.[Br-]
This item is not returnable. View return policy
14866-33-2
C32H68BrN
546.807
MFCD00011863
QBVXKDJEZKEASM-UHFFFAOYSA-M
tetraoctylammonium bromide, tetra-n-octylammonium bromide, tetraoctylazanium bromide, tetraoctyl ammonium bromide, 1-octanaminium, n,n,n-trioctyl-, bromide, unii-52qw29owoc, 52qw29owoc, tetra n-octyl ammonium bromide, acmc-1bvth, tetra-octylammonium bromide
2734117
tetraoctylazanium;bromide
CCCCCCCC[N+](CCCCCCCC)(CCCCCCCC)CCCCCCCC.[Br-]

Tetra-n-octylammonium bromide is associated with benzethonium chloride and used as a disinfectant, in surface treatment and preservative in hospitals and industries. It is used as a surfactant involved in the synthesis of blue-luminescent silicon nanoparticles in order to get uniform particle size. It is utilized as a cationic additive in the preparation of membranes using cyclam derivative 1,4,8,11-tetra(n-octyl)-1,4,8,11-tetraazacyclotetradecane as an ionphore and dibutyl phthalate as plasticizers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetra-n-octylammonium bromide is associated with benzethonium chloride and used as a disinfectant, in surface treatment and preservative in hospitals and industries. It is used as a surfactant involved in the synthesis of blue-luminescent silicon nanoparticles in order to get uniform particle size. It is utilized as a cationic additive in the preparation of membranes using cyclam derivative 1,4,8,11-tetra(n-octyl)-1,4,8,11-tetraazacyclotetradecane as an ionphore and dibutyl phthalate as plasticizers.

Solubility
Soluble in water, dichloromethane, chloroform, acetone and methanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 94°C to 99°C
Assay Percent Range 98%
Quantity 10 g
Beilstein 4169176
Solubility Information Soluble in water,dichloromethane,chloroform,acetone and methanol.
Formula Weight 546.81
Percent Purity ≥98%
Chemical Name or Material Tetra-n-octylammonium Bromide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'