missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-hexylammonium perchlorate, Thermo Scientific Chemicals

Catalog Number p-7049897
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
4656-81-9
C24H52ClNO4
454.13
MFCD00043185
RLCKPGXHGKSGOS-UHFFFAOYSA-M
tetra-n-hexylammonium perchlorate, tetrahexylammonium perchlorate, tetrahexylazanium perchlorate, 1-hexanaminium, n,n,n-trihexyl-, perchlorate, n,n,n-trihexylhexan-1-aminiumperchlorate, 1-hexanaminium, n,n,n-trihexyl-, perchlorate 1:1, tetra-n-hexyl ammonium perchlorate
78388
[O-][Cl](=O)(=O)=O.CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC
This item is not returnable. View return policy
4656-81-9
C24H52ClNO4
454.13
MFCD00043185
RLCKPGXHGKSGOS-UHFFFAOYSA-M
tetra-n-hexylammonium perchlorate, tetrahexylammonium perchlorate, tetrahexylazanium perchlorate, 1-hexanaminium, n,n,n-trihexyl-, perchlorate, n,n,n-trihexylhexan-1-aminiumperchlorate, 1-hexanaminium, n,n,n-trihexyl-, perchlorate 1:1, tetra-n-hexyl ammonium perchlorate
78388
[O-][Cl](=O)(=O)=O.CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 5 g
UN Number UN1479
IUPAC Name tetrahexylazanium;perchlorate
Formula Weight 454.13
Physical Form Crystalline
Chemical Name or Material Tetra-n-hexylammonium perchlorate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'