missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butyltin, 96%, Thermo Scientific Chemicals

Catalog Number 10785801
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1461-25-2
MFCD00009422
AFCAKJKUYFLYFK-UHFFFAOYSA-N
tetrabutyltin, tetra-n-butyltin, stannane, tetrabutyl, tin, tetrabutyl, tetra-n-butylcin, tetra-n-butylcin czech, unii-qj7y5v377v, ccris 6322, tetra-normal-butyltin, wln: 4-sn-4&4&4
15098
tetrabutylstannane
CCCC[Sn](CCCC)(CCCC)CCCC
This item is not returnable. View return policy
1461-25-2
MFCD00009422
AFCAKJKUYFLYFK-UHFFFAOYSA-N
tetrabutyltin, tetra-n-butyltin, stannane, tetrabutyl, tin, tetrabutyl, tetra-n-butylcin, tetra-n-butylcin czech, unii-qj7y5v377v, ccris 6322, tetra-normal-butyltin, wln: 4-sn-4&4&4
15098
tetrabutylstannane
CCCC[Sn](CCCC)(CCCC)CCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -97°C
Color Colorless to Straw
Density 1.0570g/mL
Boiling Point 145°C (10.0 mmHg)
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Molecular Formula C16H36Sn
Linear Formula [CH3(CH2)3]4Sn
Refractive Index 1.4725 to 1.4745
Quantity 5 g
Specific Gravity 1.057
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Molecular Weight (g/mol) 347.14
Viscosity 4 mPa.s (20°C)
Formula Weight 347.14
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Tetra-n-butyltin
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'