missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Tetra-n-butylgermanium, 99%

Brand:  Alfa Aesar™ 034119.03

Code : NEW

Additional Details : CAS Number : 1067-42-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 11347788

  • £26.30 / 1g


Estimated Shipment date
from Supplier 29-05-2018
Add to basket

Specifications

Chemical Identifiers

Tetra-n-butylgermanium
1067-42-1
301.05
MFCD00015224

Specifications

Tetra-n-butylgermanium
1067-42-1
>110° (230°F)
1.4571
99%
MFCD00015224
277° to 279°C
1g
0.934g/mL at 20°C
Liquid
301.05
Videos
SDS