missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetra-n-butylammonium sulfate, 50% w/w aq. soln., Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Description
Tetra-n-butylammonium sulfate is used as a phase transfer catalyst, neutral pH and organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Density | 1.01 |
Boiling Point | 84°C |
Odor | Unpleasant |
Refractive Index | 1.417 |
Quantity | 100 mL |
Solubility Information | Fully miscible. |
Formula Weight | 580.99 |
Concentration or Composition (by Analyte or Components) | 50% w/w aq. soln. |
Physical Form | Liquid |
Chemical Name or Material | Tetra-n-butylammonium sulfate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'