missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetra-n-butylammonium perchlorate, electrochemical grade, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
Suitable for use as a polarographic supporting electrolyteTetra-n-butylammonium perchlorate is used as a supporting electrolyte in polarographic measurements.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 210°C to 217°C |
Color | White |
Linear Formula | [CH3(CH2)3]4NClO4 |
Quantity | 10 g |
UN Number | UN1481 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in acetonitrile and ethanol; Very slightly soluble in water. |
Formula Weight | 341.92 |
Grade | Electrochemical |
Physical Form | Crystalline |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'