missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium hydroxide, 40% w/w aq. soln., Thermo Scientific Chemicals

Catalog Number p-7043801
Click to view available options
Quantity:
50 g
250 g
1000 g
2052-49-5
C16H37NO
259.48
MFCD00009425
VDZOOKBUILJEDG-UHFFFAOYSA-M
tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
2723671
[OH-].CCCC[N+](CCCC)(CCCC)CCCC
2052-49-5
C16H37NO
259.48
MFCD00009425
VDZOOKBUILJEDG-UHFFFAOYSA-M
tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
2723671
[OH-].CCCC[N+](CCCC)(CCCC)CCCC

Tetra-n-butylammonium hydroxide, 40% w/w aq. solution is used as a phase-transfer catalyst in organic synthesis especially for alkylation and condensation reactions. It is used as a curing accelerator for epoxy resins. It is also used in preparation of dichlorocarbene from chloroform.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetra-n-butylammonium hydroxide, 40% w/w aq. solution is used as a phase-transfer catalyst in organic synthesis especially for alkylation and condensation reactions. It is used as a curing accelerator for epoxy resins. It is also used in preparation of dichlorocarbene from chloroform.

Solubility
Miscible with organic solvents.

Notes
Air sensitive and hygroscopic. Incompatible with strong acids and corrodes metal.
TRUSTED_SUSTAINABILITY
Density 0.99
pH 14.0
Flash Point None
Odor Amine-like
Linear Formula [CH3(CH2)3]4NOH
Refractive Index 1.408
Quantity 50 g
UN Number UN3267
Beilstein 3571228
Sensitivity Air Sensitive
Solubility Information Miscible with organic solvents.
IUPAC Name tetrabutylazanium hydroxide
Formula Weight 259.48
Concentration or Composition (by Analyte or Components) 40% w/w aq. soln.
Chemical Name or Material Tetra-n-butylammonium hydroxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'