missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetra-n-butylammonium hydroxide, 1.0M aq. soln., HPLC grade, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Description
Phase transfer catalyst, electrolyte in polarography, ion-pair extractionTetra-n-butylammonium hydroxide, 1.0M in aqueous solution is used as a strong base in organic chemistry. It is utilized under phase-transfer conditions to effect alkylation and deprotonation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Flash Point | None |
Linear Formula | [CH3(CH2)3]4NOH |
Quantity | 100 mL |
UN Number | UN3267 |
Beilstein | 3571228 |
Sensitivity | Air Sensitive |
Solubility Information | Miscible with organic solvents. |
Formula Weight | 259.48 |
Grade | HPLC |
Concentration or Composition (by Analyte or Components) | 1.0M aq. soln. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'