missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetra-n-butylammonium hexafluorophosphate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Tetra-n-butylammonium hexafluorophosphate is used as a supporting electrolyte in nonaqueous electrochemistry. It is chemically inert, thereby allowing the usage as an inert electrolyte with potential range. It enhances the performance of single-layer organic light emitting diodes (OLEDs).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tetra-n-butylammonium hexafluorophosphate is used as a supporting electrolyte in nonaqueous electrochemistry. It is chemically inert, thereby allowing the usage as an inert electrolyte with potential range. It enhances the performance of single-layer organic light emitting diodes (OLEDs).
Solubility
Soluble in acetone, acetonitrile and tetrhydrofuran.
Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Tetra-n-butylammonium hexafluorophosphate is used as a supporting electrolyte in nonaqueous electrochemistry. It is chemically inert, thereby allowing the usage as an inert electrolyte with potential range. It enhances the performance of single-layer organic light emitting diodes (OLEDs).
Solubility
Soluble in acetone, acetonitrile and tetrhydrofuran.
Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
Melting Point | 242°C to 248°C |
Linear Formula | [CH3(CH2)3]4NPF6 |
Quantity | 25 g |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in acetone,acetonitrile and tetrhydrofuran. |
IUPAC Name | tetrabutylazanium;hexafluorophosphate |
Formula Weight | 387.44 |
Percent Purity | 98% |
Chemical Name or Material | Tetra-n-butylammonium hexafluorophosphate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'