missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetra-n-butylammonium acetate, Thermo Scientific Chemicals
23.20 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Description
Tetra-n-butylammonium acetate is used as catalyst, curing agent, and initiator in the polymer industry. It is also used catalyst for the alkynylation of carbonyl compounds with trimethylsilylacetylenes. It is also used as PE/PP clarifying agent, epoxy polymerization initiator, surfactants, emulsifiers, and antistatic agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 114°C to 115°C |
Color | White |
Linear Formula | [CH3(CH2)3]4N(O2CCH3) |
Quantity | 1 g |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in acetonitrile (0.1g/mL). |
Formula Weight | 301.51 |
Physical Form | Crystalline |
Chemical Name or Material | Tetra-n-butylammonium acetate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'