missing translation for 'onlineSavingsMsg'
Learn More

Tetra-N-Butyl Ammonium Hydroxide Solution, 40%, Extra Pure, SLR, Fisher Chemical™

Catalog Number p-7073600
Click to view available options
Quantity:
250 mL
This item is not returnable. View return policy
2052-49-5
C16H37NO
259.48
MFCD00009425
VDZOOKBUILJEDG-UHFFFAOYSA-M
tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
2723671
tetrabutylazanium hydroxide
[OH-].CCCC[N+](CCCC)(CCCC)CCCC
This item is not returnable. View return policy
2052-49-5
C16H37NO
259.48
MFCD00009425
VDZOOKBUILJEDG-UHFFFAOYSA-M
tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
2723671
tetrabutylazanium hydroxide
[OH-].CCCC[N+](CCCC)(CCCC)CCCC
Color Yellow
pH 14
Physical Form Liquid
Quantity 250 mL
Formula Weight 259.46g/mol
Packaging HDPE plastic bottle
compliance-icons
Product Identifier
  • Tetrabutylammonium hydroxide, 40 wt.% solution in water
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'