missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetra-N-Butyl Ammonium Hydroxide Solution 0.1M (0.1N) In Toluene/Methanol, Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
255.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
255.26 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "22914" to get your promotional price.
255.26 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 L
Packaging:
Amber glass bottle
Bag in box
Description
Tetra-N-Butyl Ammonium Hydroxide Solution 0.1M (0.1N) In Toluene/Methanol, Standard Solution Ready To Use, for Volumetric Analysis
Specifications
Color | Colorless |
Physical Form | Liquid |
Quantity | 1 L |
Formula Weight | 259.47g/mol |
Packaging | Amber glass bottle |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'