missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetra-N-Butyl Ammonium Hydroxide Solution 0.1M (0.1N) In Toluene/Methanol, Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 L
Packaging:
Amber glass bottle
Bag in box
Description
Tetra-N-Butyl Ammonium Hydroxide Solution 0.1M (0.1N) In Toluene/Methanol, Standard Solution Ready To Use, for Volumetric Analysis
Specifications
Color | Colorless |
Physical Form | Liquid |
Quantity | 2.5 L |
Formula Weight | 259.47g/mol |
Packaging | Amber glass bottle |
Safety and Handling
Product Identifier
- Tetrabutylammonium hydroxide, 0.1N sol. in toluene/methanol
- Danger
- Acute toxicity Category 3
- Aspiration toxicity Category 1
- Specific target organ toxicity Category 3
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after single exposure Category 1
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 4
- H304-May be fatal if swallowed and enters airways.
- H370-Causes damage to organs.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H225-Highly flammable liquid and vapour.
- H336-May cause drowsiness or dizziness.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P331-Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'