Learn More
R&D Systems™ Testosterone Parameter Assay Kit, R&D Systems™
Testosterone Parameter Assay Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand:  R&D Systems™ KGE010
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13438055
  • £370.93 / 1 set
Estimated Shipment
28-06-2019
Description
Description
The Testosterone Parameter Assay Kit quantifies multi-species Testosterone in cell culture supernates (100 ul), serum (20 ul), edta plasma (20 ul), heparin plasma (20 ul).
Specifications
Specifications
0.04 - 10ng/mL
Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules.
ELISA kit
0.041ng/mL
Multi-species
1 x 96-well plate
Biotin
ELISA
1 kit
Cell Culture Supernates (100uL), Serum (20uL), EDTA Plasma (20uL), Heparin Plasma (20uL)
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Videos