missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Testosterone Parameter Assay Kit, R&D Systems™
Testosterone Parameter Assay Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
POA
Specifications
Assay Range | 0.04 - 10ng/mL |
---|---|
Assay Sensitivity | 0.041ng/mL |
Sample Type | Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum |
Description
The Testosterone Parameter Assay Kit quantifies multi-species Testosterone in cell culture supernates (100 ul), serum (20 ul), edta plasma (20 ul), heparin plasma (20 ul).Specifications
0.04 - 10ng/mL | |
0.041ng/mL | |
Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum |