missing translation for 'onlineSavingsMsg'
Learn More

Testosterone, Eur.Pharm., Thermo Scientific Chemicals

Catalog Number p-7214334
Click to view available options
Quantity:
5 g
10 g
25 g
This item is not returnable. View return policy
Regulatory: The product is controlled by Misuse of Drugs Act 1971 and the Misuse of Drugs Regulations 2001, all customers must complete a declaration before delivery.
58-22-0
C19H28O2
288.43
MUMGGOZAMZWBJJ-DYKIIFRCSA-N
(1S,3aS,3bR,9aR,9bS,11aS)-1-hydroxy-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2O
This item is not returnable. View return policy
Regulatory: The product is controlled by Misuse of Drugs Act 1971 and the Misuse of Drugs Regulations 2001, all customers must complete a declaration before delivery.
58-22-0
C19H28O2
288.43
MUMGGOZAMZWBJJ-DYKIIFRCSA-N
(1S,3aS,3bR,9aR,9bS,11aS)-1-hydroxy-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2O

Testosterone, CAS # 58-22-0, is a steroid sex hormone from the androstane family and it is important to regulate and maintain several biological functions.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Testosterone is a steroid sex hormone from the androstane family and is important for the maintenance of several biological functions

• This compound is an androgen that exerts its function by binding to and activating the androgen receptor

Applications

• Testosterone is irreversibly converted to dihydrotestosterone (DHT) in target tissues by the enzyme 5-∝ reductase

• Testosterone or DHT (its active metabolite) ligand-androgen receptor complexes act as a transcription factor, stimulating the expression of several responsive genes

• It antagonizes the androgen receptor to induce gene expression that causes the growth, development, and maintenance of several biological functions

• This compound is commonly used in vivo studies as an ester derivative due to its rapid metabolism by the liver

TRUSTED_SUSTAINABILITY
Melting Point 153.0°C to 157.0°C
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 9324
Specific Rotation +110.00
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: soluble in chloroform, alcohol, acetone, benzene,, dioxane and vegetable oils, slightly soluble in ether
Formula Weight 288.42
Chemical Name or Material Testosterone
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Testosterone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 2
  • Reproductive toxicity on or via lactation
  • Reproductive toxicity Category 1A
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H360FD-May damage fertility. May damage the unborn child.
  • H351-Suspected of causing cancer.
  • H362-May cause harm to breast-fed children.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'