missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Karl Hecht™ Test Tubes Borosilicate Glass With Beaded Rim

Manufactured from borosilicate glass. DURAN™ Karl Hecht™ Test Tubes Borosilicate Glass With Beaded Rim features good thermal shock resistance.

Brand:  Duran™ 261300105

Code : A0

Additional Details : Weight : 0.30000kg

 View more versions of this product

Product Code. 10613721

  • £40.65 / Pack of 100


Estimated Shipment
14-02-2018
Add to basket

Description

Description

  • Beaded rim
  • Good thermal shock resistance
  • Manufactured from borosilicate glass
Specifications

Specifications

Yes
Suited to Rapid Temperature Changes or Localized Heating
Yes
Borosilicate Glass
8mm
100Pack
2mL
Test Tube
Videos