Learn More
Palintest™ Test Chlorine DPD No 1 + No 3
With interchangeable colour discs
Brand:  Palintest™ CM 031/1
Code : 04
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 13181521
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
The Palintest™ comparator with interchangeable color discs is simple to use. Just add the reagent tablet to the test sample, place in the comparator and match against the appropriate disc.
Specifications
Specifications
50/Pk.
50/Pk.