missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Nitrosobutane dimer, 98+%, Thermo Scientific Chemicals
21.53 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 mg
1 g
Description
tert-Nitrosobutane dimer is used as a spin trapping reagent in electron spin resonance studies. Spin trap reagent, Reactant involved in the synthesis of air-stable π-radical based on Corannulene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
tert-Nitrosobutane dimer is used as a spin trapping reagent in electron spin resonance studies. Spin trap reagent, Reactant involved in the synthesis of air-stable π-radical based on Corannulene.
Solubility
Soluble in 5% chloroform.
Notes
Light Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
tert-Nitrosobutane dimer is used as a spin trapping reagent in electron spin resonance studies. Spin trap reagent, Reactant involved in the synthesis of air-stable π-radical based on Corannulene.
Solubility
Soluble in 5% chloroform.
Notes
Light Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | 88°C to 92°C (sublimation) |
Quantity | 250 mg |
Sensitivity | Light sensitive |
Solubility Information | Soluble in 5% chloroform. |
Formula Weight | 174.24 |
Percent Purity | ≥98% |
Chemical Name or Material | tert-Nitrosobutane dimer |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'