missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylperoxy 2-ethylhexyl carbonate, 95%, Acros Organics

Brand:  Acros Organics 349850100

Code : 29

Additional Details : CAS Number : 34443-12-4 Weight : 0.05930kg Transport : UN number : 3105 Chem class : 12 Pack group : 0


Quantity 10mL

 View more versions of this product

Product Code. 10788342

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

tert-Butylperoxy 2-ethylhexyl carbonate
95%
C13H26O4
246.35

Specifications

tert-Butylperoxy 2-ethylhexyl carbonate
95%
Plastic bottle
10mL
34443-12-4
C13H26O4
246.35
Videos
SDS