missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ tert-Butylferrocene, 98%

Brand:  Alfa Aesar™ 039405.06

Code : NEW

Additional Details : CAS Number : 1316-98-9 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11378188

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

tert-Butylferrocene
1316-98-9
242.15
MFCD00058729

Specifications

tert-Butylferrocene
1.201g/mL
98%
MFCD00058729
96°C (1mmHg)
1316-98-9
Liquid
242.15
1.5790
5g
Videos
SDS