missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyldimethylsilyl trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

Catalog Number p-7081964
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
69739-34-0
C7H15F3O3SSi
264.33
MFCD00000405
WLLIXJBWWFGEHT-UHFFFAOYSA-N
tert-butyldimethylsilyl trifluoromethanesulfonate, trifluoromethanesulfonic acid tert-butyldimethylsilyl ester, tbdms triflate, t-butyldimethylsilyl triflate, tert-butyldimethylsilyl trifluoromethanesulphonate, tbdms-otf, tert-butyldimethylsilyl triflate, silane tbm2 triflate, t-butyldimethylsilyltrifluoromethanesulfonate, t-butylmethylsilyl triflate
123564
[tert-butyl(dimethyl)silyl] trifluoromethanesulfonate
CC(C)(C)[Si](C)(C)OS(=O)(=O)C(F)(F)F
This item is not returnable. View return policy
69739-34-0
C7H15F3O3SSi
264.33
MFCD00000405
WLLIXJBWWFGEHT-UHFFFAOYSA-N
tert-butyldimethylsilyl trifluoromethanesulfonate, trifluoromethanesulfonic acid tert-butyldimethylsilyl ester, tbdms triflate, t-butyldimethylsilyl triflate, tert-butyldimethylsilyl trifluoromethanesulphonate, tbdms-otf, tert-butyldimethylsilyl triflate, silane tbm2 triflate, t-butyldimethylsilyltrifluoromethanesulfonate, t-butylmethylsilyl triflate
123564
[tert-butyl(dimethyl)silyl] trifluoromethanesulfonate
CC(C)(C)[Si](C)(C)OS(=O)(=O)C(F)(F)F

tert-Butyldimethylsilyl trifluoromethanesulfonate is involved in the introduction of a bulky tert-butyl dimethylsilyl group onto a cis-bis(alkenyl)oxirane used in Cope rearrangement. It is associated with a thiolane and promotes the chalcogenide-Morita-Baylis-Hillman reaction. Further, it is used as a highly reactive silylating agent. In addition to this, it is used to prepare enol silyl ethers by reacting with ketones and lactones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyldimethylsilyl trifluoromethanesulfonate is involved in the introduction of a bulky tert-butyl dimethylsilyl group onto a cis-bis(alkenyl)oxirane used in Cope rearrangement. It is associated with a thiolane and promotes the chalcogenide-Morita-Baylis-Hillman reaction. Further, it is used as a highly reactive silylating agent. In addition to this, it is used to prepare enol silyl ethers by reacting with ketones and lactones.

Solubility
Slightly miscible with chloroform.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong acids.
TRUSTED_SUSTAINABILITY
Density 1.151
Boiling Point 65°C to 67°C (12 mmHg)
Flash Point 36°C (96°F)
Refractive Index 1.386
Quantity 5 g
UN Number UN2920
Beilstein 2370068
Sensitivity Moisture sensitive
Solubility Information Slightly miscible with chloroform.
Formula Weight 264.34
Percent Purity 98%
Chemical Name or Material tert-Butyldimethylsilyl trifluoromethanesulfonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'