missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyldimethylsilane, 95%, Thermo Scientific Chemicals

Catalog Number p-4380060
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
29681-57-0
C6H16Si
116.28
ILMRJRBKQSSXGY-UHFFFAOYSA-N
tert-butyldimethylsilane, t-butyldimethylsilane, tert-butyldimethylsilyl, silane, 1,1-dimethylethyl dimethyl, t-butyldimethylsilyl, t-butyl-dimethylsilyl, t-butyl-dimethyl-silyl, tert-butyl dimethylsilane, tert-butyldimethylsilyl group
9898815
tert-butyl(dimethyl)silicon
CC(C)(C)[Si](C)C
This item is not returnable. View return policy
29681-57-0
C6H16Si
116.28
ILMRJRBKQSSXGY-UHFFFAOYSA-N
tert-butyldimethylsilane, t-butyldimethylsilane, tert-butyldimethylsilyl, silane, 1,1-dimethylethyl dimethyl, t-butyldimethylsilyl, t-butyl-dimethylsilyl, t-butyl-dimethyl-silyl, tert-butyl dimethylsilane, tert-butyldimethylsilyl group
9898815
tert-butyl(dimethyl)silicon
CC(C)(C)[Si](C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.7010g/mL
Boiling Point 81°C to 83°C
Flash Point -11°C
Assay Percent Range 94% min. (GC)
Quantity 5 g
Specific Gravity 0.701
Formula Weight 116.28
Percent Purity 95%
Chemical Name or Material tert-Butyldimethylsilane
compliance-icons compliance-icons
Product Identifier
  • tert-Butyldimethylsilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'