missing translation for 'onlineSavingsMsg'
Learn More

tert-butyl6-bromo-4-oxo-3,4-dihydro-1'H-spiro[1,3-benzoxazine-2,4'-piperidine]-1'-, Thermo Scientific™

Catalog Number 10451453
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
690632-05-4
C17H21BrN2O4
397.269
MFCD05865135
FKBBJWUNJKLMBK-UHFFFAOYSA-N
tert-butyl 6-bromo-4-oxo-3,4-dihydro-1'h-spiro 1,3-benzoxazine-2,4'-piperidine-1'-carboxylate, tert-butyl 6-bromo-4-oxo-3,4-dihydrospiro benzo e 1,3 oxazine-2,4'-piperidine-1'-carboxylate, 6-bromospiro 1,3-benzoxazine-2,4'-piperidine-4 3h-one, n-boc protected, tert-butyl 6-bromo-4-oxo-3h-spiro 1,3-benzoxazine-2,4'-piperidine-1'-carboxylate, tert-butyl 6-bromo-4-oxospiro 3h-1,3-benzoxazine-2,4'-piperidine-1'-carboxylate, 6-bromo-3,4-dihydro-4-oxo-spiro 2h-1,3-benzoxazine-2,4'-piperidin-1'-carboxylic acid tert-butyl ester, spiro 2h-1,3-benzoxazine-2,4'-piperidine-1'-carboxylicacid,6-bromo-3,4-dihydro-4-oxo-,1,1-dimethylethyl ester, tert-butyl 6-bromo-3,4-dihydro-4-oxo-1'h-spiro 1,3-benzoxazine-2,4'-piperidine-1'-carboxylate
2794762
tert-butyl 6-bromo-4-oxospiro[3H-1,3-benzoxazine-2,4'-piperidine]-1'-carboxylate
CC(C)(C)OC(=O)N1CCC2(CC1)NC(=O)C3=C(O2)C=CC(=C3)Br
This item is not returnable. View return policy
690632-05-4
C17H21BrN2O4
397.269
MFCD05865135
FKBBJWUNJKLMBK-UHFFFAOYSA-N
tert-butyl 6-bromo-4-oxo-3,4-dihydro-1'h-spiro 1,3-benzoxazine-2,4'-piperidine-1'-carboxylate, tert-butyl 6-bromo-4-oxo-3,4-dihydrospiro benzo e 1,3 oxazine-2,4'-piperidine-1'-carboxylate, 6-bromospiro 1,3-benzoxazine-2,4'-piperidine-4 3h-one, n-boc protected, tert-butyl 6-bromo-4-oxo-3h-spiro 1,3-benzoxazine-2,4'-piperidine-1'-carboxylate, tert-butyl 6-bromo-4-oxospiro 3h-1,3-benzoxazine-2,4'-piperidine-1'-carboxylate, 6-bromo-3,4-dihydro-4-oxo-spiro 2h-1,3-benzoxazine-2,4'-piperidin-1'-carboxylic acid tert-butyl ester, spiro 2h-1,3-benzoxazine-2,4'-piperidine-1'-carboxylicacid,6-bromo-3,4-dihydro-4-oxo-,1,1-dimethylethyl ester, tert-butyl 6-bromo-3,4-dihydro-4-oxo-1'h-spiro 1,3-benzoxazine-2,4'-piperidine-1'-carboxylate
2794762
tert-butyl 6-bromo-4-oxospiro[3H-1,3-benzoxazine-2,4'-piperidine]-1'-carboxylate
CC(C)(C)OC(=O)N1CCC2(CC1)NC(=O)C3=C(O2)C=CC(=C3)Br
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 397.27
Percent Purity 95%
Chemical Name or Material tert-Butyl 6-bromo-4-oxo-3,4-dihydro-1';H-spiro[1,3-benzoxazine-2,4';-piperidine]-1';-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'