missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-butyl (S)-4-(dimethylamino)-1-(piperazine-1-carbonyl)butylcarbamate, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 5 g |
Formula Weight | 328.45 |
Percent Purity | 97% |
Chemical Name or Material | tert-Butyl (S)-4-(dimethylamino)-1-(piperazine-1-carbonyl)butylcarbamate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'