missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ tert-Butyl (S)-2-aminobutyrate hydrochloride, 95%

Brand:  Alfa Aesar™ H65284.06

Code : NEW

Additional Details : CAS Number : 53956-05-1 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15412509

  • £63.20 / 5g


Estimated Shipment date
from Supplier 07-02-2018
Add to basket

Specifications

Chemical Identifiers

tert-Butyl (S)-2-aminobutyrate hydrochloride
195.69
6935251
53956-05-1
MFCD00237697
H-2-Abu-OtBu.HCl; (S)-2-Aminobutyric acid tert-butyl ester hydrochloride

Specifications

tert-Butyl (S)-2-aminobutyrate hydrochloride
Solid
195.69
6935251
5g
53956-05-1
95%
MFCD00237697
H-2-Abu-OtBu.HCl; (S)-2-Aminobutyric acid tert-butyl ester hydrochloride
Videos
SDS