missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ tert-Butyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate, 95%

Brand:  Alfa Aesar™ H62911.03

Code : NEW

Additional Details : CAS Number : 91229-91-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15427996

  • £147.34 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

tert-Butyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate
285.34
Boc-L-Pyr-Otbu; (S)-N-Boc-pyroglutamic acid tert-butyl ester
91229-91-3
MFCD03094770

Specifications

tert-Butyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate
Powder
285.34
Boc-L-Pyr-Otbu; (S)-N-Boc-pyroglutamic acid tert-butyl ester
1g
91229-91-3
95%
MFCD03094770
White
Videos
SDS