missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl propargyl ether, 97%, Thermo Scientific Chemicals

Catalog Number 10679204
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
16314-18-4
C7H12O
112.17
MFCD07368651
QIGSPXPODKLGRK-UHFFFAOYSA-N
7172302
CC(C)(C)OCC#C
This item is not returnable. View return policy
16314-18-4
C7H12O
112.17
MFCD07368651
QIGSPXPODKLGRK-UHFFFAOYSA-N
7172302
CC(C)(C)OCC#C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow
Density 0.8350g/mL
Boiling Point 119°C
Flash Point 20°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH∽CCH2OC(CH3)3
Refractive Index 1.4170 to 1.419
Quantity 1 g
Specific Gravity 0.835
Synonym tert-butyl propargyl ether, 3-tert-butoxy prop-1-yne, 1-propyne,3-1,1-dimethylethoxy, 1-propyne, 3-1,1-dimethylethoxy, 3-tert-butoxyprop-1-yne, 3-tert-butyloxy-1-propyne, tert-butylpropargylether
IUPAC Name 2-methyl-2-prop-2-ynoxypropane
Formula Weight 112.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material tert-Butyl propargyl ether
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • tert-Butyl propargyl ether
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'