missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl N-[4-(aminomethyl)phenyl]carbamate, 97%, Thermo Scientific™

Catalog Number p-3111190
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy
220298-96-4
C12H18N2O2
222.29
MFCD02183573
URXUHALBOWYXJZ-UHFFFAOYSA-N
tert-butyl n-4-aminomethyl phenyl carbamate, 4-aminomethyl-1-n-boc-aniline, tert-butyl 4-aminomethyl phenyl carbamate, 4-boc-amino benzylamine, 4-aminomethy-1-n-boc-aniline, 4-n-boc-amino benzylamine, 4-tert-butoxycarbonylamino benzylamine, 4-aminomethyl-n-boc-aniline, tert-butyl-n-4-aminomethyl phenyl carbamate
2794659
tert-butyl N-[4-(aminomethyl)phenyl]carbamate
CC(C)(C)OC(=O)NC1=CC=C(CN)C=C1
This item is not returnable. View return policy
220298-96-4
C12H18N2O2
222.29
MFCD02183573
URXUHALBOWYXJZ-UHFFFAOYSA-N
tert-butyl n-4-aminomethyl phenyl carbamate, 4-aminomethyl-1-n-boc-aniline, tert-butyl 4-aminomethyl phenyl carbamate, 4-boc-amino benzylamine, 4-aminomethy-1-n-boc-aniline, 4-n-boc-amino benzylamine, 4-tert-butoxycarbonylamino benzylamine, 4-aminomethyl-n-boc-aniline, tert-butyl-n-4-aminomethyl phenyl carbamate
2794659
tert-butyl N-[4-(aminomethyl)phenyl]carbamate
CC(C)(C)OC(=O)NC1=CC=C(CN)C=C1
CAS Min % 97.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 222.29
Percent Purity 97%
Chemical Name or Material tert-Butyl N-[4-(aminomethyl)phenyl]carbamate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'