Learn More
Thermo Scientific Maybridge tert-Butyl N-(2-thienyl)carbamate, 1g, 97%
Brand: Thermo Scientific Maybridge CC00045DA
tert-Butyl N-(2-thienyl)carbamate, 1g, 97%
Code : 31
Additional Details : CAS Number : 56267-50-6 Weight : 0.05100kg
Brand: Thermo Scientific Maybridge CC00045DA
tert-Butyl N-(2-thienyl)carbamate, 1g, 97%
Code : 31
Additional Details : CAS Number : 56267-50-6 Weight : 0.05100kg