missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl N-[(1-benzyl-4-piperidinyl)methyl]carbamate, ≥95%, Thermo Scientific™

Catalog Number p-3076460
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
173340-23-3
C18H28N2O2
304.434
MFCD04123925
SOXSDIKJHXTJEP-UHFFFAOYSA-N
tert-butyl 1-benzylpiperidin-4-yl methyl carbamate, tert-butyl 1-benzylpiperidin-4-yl methylcarbamate, tert-butyl n-1-benzyl-4-piperidinyl methyl carbamate, tert-butyl n-1-benzylpiperidin-4-yl methyl carbamate, maybridge3_004457, 1-benzyl-4-tertbutyloxycarbonylaminomethylpiperidine, 1-benzyl-4-piperidinylmethyl carbamic acid tert-butyl ester, 1-benzylpiperidin-4-yl methylcarbamic acid tert-butyl ester, carbamic acid,n-1-phenylmethyl-4-piperidinyl methyl-, 1,1-dimethylethyl ester
2824066
tert-butyl N-[(1-benzylpiperidin-4-yl)methyl]carbamate
CC(C)(C)OC(=O)NCC1CCN(CC1)CC2=CC=CC=C2
This item is not returnable. View return policy
173340-23-3
C18H28N2O2
304.434
MFCD04123925
SOXSDIKJHXTJEP-UHFFFAOYSA-N
tert-butyl 1-benzylpiperidin-4-yl methyl carbamate, tert-butyl 1-benzylpiperidin-4-yl methylcarbamate, tert-butyl n-1-benzyl-4-piperidinyl methyl carbamate, tert-butyl n-1-benzylpiperidin-4-yl methyl carbamate, maybridge3_004457, 1-benzyl-4-tertbutyloxycarbonylaminomethylpiperidine, 1-benzyl-4-piperidinylmethyl carbamic acid tert-butyl ester, 1-benzylpiperidin-4-yl methylcarbamic acid tert-butyl ester, carbamic acid,n-1-phenylmethyl-4-piperidinyl methyl-, 1,1-dimethylethyl ester
2824066
tert-butyl N-[(1-benzylpiperidin-4-yl)methyl]carbamate
CC(C)(C)OC(=O)NCC1CCN(CC1)CC2=CC=CC=C2
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 304.43
Percent Purity 95%
Chemical Name or Material tert-Butyl N-[(1-benzyl-4-piperidinyl)methyl]carbamate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'