missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl methyl ether, ACS Reagent, ≥99%, Honeywell™ Riedel-de Haën™

Catalog Number p-7120861
Click to view available options
Quantity:
500 mL
1 L
2 L
2.5 L
25 L
4 x 2.5 L
Packaging:
Glass Bottle
Stainless Steel Can
This item is not returnable. View return policy
1634-04-4
C5H12O
88.15
MFCD00008812
BZLVMXJERCGZMT-UHFFFAOYSA-N
tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
15413
CHEBI:27642
2-methoxy-2-methylpropane
CC(C)(C)OC
This item is not returnable. View return policy
1634-04-4
C5H12O
88.15
MFCD00008812
BZLVMXJERCGZMT-UHFFFAOYSA-N
tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
15413
CHEBI:27642
2-methoxy-2-methylpropane
CC(C)(C)OC

ACS Reagent,≥99.0%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY
Melting Point -109°C
Linear Formula (CH3)3COCH3
UN Number UN2398
Beilstein 1730942
Solubility Information In water, 42g/L at 20°C
Formula Weight 88.15g/mol
Percent Purity ≥99%
Density 0.74g/mL at 25°C
Boiling Point 55.3°C
Quantity 500 mL
Explosion Limit 0.151
Flash Point −33°C (−27.4°F)
Ignition Point 705°F
Odor Aromatic
Packaging Glass Bottle
Physical Form Liquid
Chemical Name or Material tert-Butyl methyl ether
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'