missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl hydrogen malonate, 96%, Thermo Scientific Chemicals

Catalog Number p-7028814
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
40052-13-9
C7H12O4
160.169
MFCD00191886
NGGGZUAEOKRHMA-UHFFFAOYSA-N
3-tert-butoxy-3-oxopropanoic acid, malonic acid mono-tert-butyl ester, mono-tert-butyl malonate, 2-tert-butoxycarbonyl acetic acid, tert-butyl hydrogen malonate, mono-t-butyl malonate, pubchem13754, malonicacidmono-tert-butylester, malonic acid mono t-butyl ester
545853
3-[(2-methylpropan-2-yl)oxy]-3-oxopropanoic acid
CC(C)(C)OC(=O)CC(=O)O
This item is not returnable. View return policy
40052-13-9
C7H12O4
160.169
MFCD00191886
NGGGZUAEOKRHMA-UHFFFAOYSA-N
3-tert-butoxy-3-oxopropanoic acid, malonic acid mono-tert-butyl ester, mono-tert-butyl malonate, 2-tert-butoxycarbonyl acetic acid, tert-butyl hydrogen malonate, mono-t-butyl malonate, pubchem13754, malonicacidmono-tert-butylester, malonic acid mono t-butyl ester
545853
3-[(2-methylpropan-2-yl)oxy]-3-oxopropanoic acid
CC(C)(C)OC(=O)CC(=O)O

tert-Butyl hydrogen malonate may be used in the preparation of dendritic precursor to asymmetric methanofullerenes, hapten-3,6-(O,S-dimethylthiophosphoramido)-6-oxohexanoic acid, hapten-4,3-(O,S-dimethylthiophosphoramido)-3-oxopropanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl hydrogen malonate may be used in the preparation of dendritic precursor to asymmetric methanofullerenes, hapten-3,6-(O,S-dimethylthiophosphoramido)-6-oxohexanoic acid, hapten-4,3-(O,S-dimethylthiophosphoramido)-3-oxopropanoic acid.

Solubility
Soluble in ethanol.

Notes
Store in a cool, dry condition in a well sealed container.
TRUSTED_SUSTAINABILITY
Melting Point 19°C to 20°C
Density 1.04
Boiling Point 90°C (2 mmHg)
Flash Point 91°C (195°F)
Refractive Index 1.426
Quantity 5 g
Beilstein 1765457
Solubility Information Soluble in ethanol.
Formula Weight 160.17
Percent Purity 96%
Chemical Name or Material tert-Butyl hydrogen malonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'