missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Ethyl Ether, 97%, Thermo Scientific Chemicals

Catalog Number 10566782
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
637-92-3
C6H14O
102.18
MFCD00009225
NUMQCACRALPSHD-UHFFFAOYSA-N
tert-butyl ethyl ether, ethyl tert-butyl ether, etbe, propane, 2-ethoxy-2-methyl, ethyl tert-butyl oxide, ether, tert-butyl ethyl, 2-methyl-2-ethoxypropane, ethyl t-butyl ether, tert-butylethylether, methyl-2-ethoxypropane
12512
2-ethoxy-2-methylpropane
CCOC(C)(C)C
637-92-3
C6H14O
102.18
MFCD00009225
NUMQCACRALPSHD-UHFFFAOYSA-N
tert-butyl ethyl ether, ethyl tert-butyl ether, etbe, propane, 2-ethoxy-2-methyl, ethyl tert-butyl oxide, ether, tert-butyl ethyl, 2-methyl-2-ethoxypropane, ethyl t-butyl ether, tert-butylethylether, methyl-2-ethoxypropane
12512
2-ethoxy-2-methylpropane
CCOC(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -94.0°C
Color Colorless
Density 0.7500g/mL
Boiling Point 71.0°C
Flash Point −19°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3COC2H5
Refractive Index 1.3746 to 1.3766
Quantity 25 g
Merck Index 15, 3829
Specific Gravity 0.75
Solubility Information Solubility in water: 1.2 g/100mL (20°C).
Formula Weight 102.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material tert-Butyl ethyl ether
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'