missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl carbamate, 98%, Thermo Scientific Chemicals

Catalog Number 10454643
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
4248-19-5
C5H11NO2
117.15
LFKDJXLFVYVEFG-UHFFFAOYSA-N
t-butyl carbamate, carbamic acid tert-butyl ester, boc amine, boc-amide, carbamic acid, 1,1-dimethylethyl ester, boc-nh2, tert-butylcarbamat, n-boc amine, boc-amine, tert butyl carbamate
77922
tert-butyl carbamate
CC(C)(C)OC(=O)N
This item is not returnable. View return policy
4248-19-5
C5H11NO2
117.15
LFKDJXLFVYVEFG-UHFFFAOYSA-N
t-butyl carbamate, carbamic acid tert-butyl ester, boc amine, boc-amide, carbamic acid, 1,1-dimethylethyl ester, boc-nh2, tert-butylcarbamat, n-boc amine, boc-amine, tert butyl carbamate
77922
tert-butyl carbamate
CC(C)(C)OC(=O)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 105.0°C to 109.0°C
Color White to Yellow
Assay Percent Range 97.5% min. (GC)
Infrared Spectrum Authentic
Linear Formula H2NCO2C(CH3)3
Beilstein 03,II,26
Packaging Glass Bottle
Solubility Information Solubility in water: insoluble
Formula Weight 117.15
Percent Purity 98%
Quantity 1 g
Physical Form Needles
Chemical Name or Material tert-Butyl carbamate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'