missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl bromide, 98+%, stab. with potassium carbonate, Thermo Scientific Chemicals

Catalog Number p-7021408
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
507-19-7
C4H9Br
137.02
MFCD00000125
RKSOPLXZQNSWAS-UHFFFAOYSA-N
tert-butyl bromide, t-butyl bromide, trimethylbromomethane, propane, 2-bromo-2-methyl, bromotrimethylmethane, 2-bromoisobutane, tertiarybutyl bromide, tert-butylbromide, 2-methyl-2-bromopropane, 1-bromo-1,1-dimethylethane
10485
2-bromo-2-methylpropane
CC(C)(C)Br
This item is not returnable. View return policy
507-19-7
C4H9Br
137.02
MFCD00000125
RKSOPLXZQNSWAS-UHFFFAOYSA-N
tert-butyl bromide, t-butyl bromide, trimethylbromomethane, propane, 2-bromo-2-methyl, bromotrimethylmethane, 2-bromoisobutane, tertiarybutyl bromide, tert-butylbromide, 2-methyl-2-bromopropane, 1-bromo-1,1-dimethylethane
10485
2-bromo-2-methylpropane
CC(C)(C)Br

tert-Butyl bromide is used as a raw material in synthetic organic chemistry. It is used to study the massive deadenylation of adenine based-nucleosides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl bromide is used as a raw material in synthetic organic chemistry. It is used to study the massive deadenylation of adenine based-nucleosides.

Solubility
Immiscible with water.

Notes
Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point °C to 20°C
Density 1.210
Boiling Point 74°C to 76°C
Flash Point 18°C (64°F)
Assay Percent Range 98%, stabilized with potassium carbonate
Refractive Index 1.428
Quantity 100 g
UN Number UN2342
Beilstein 1730892
Merck Index 14,1555
Solubility Information Immiscible with water.
Formula Weight 137.02
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material tert-Butyl Bromide, stabilized with potassium carbonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'