missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl 5-bromo-1H-indazole-1-carboxylate, ≥90%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 89.5 |
CAS Max % | 100.0 |
Assay Percent Range | 90% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | tert-butyl 5-bromo-1h-indazole-1-carboxylate, 1-boc-5-bromo-indazole, n-boc-5-bromo-1h-indazole, n-boc-5-bromoindazole, 1-boc-5-bromo-1h-indazole, n-boc-5-bromo indazole, 1h-indazole-1-carboxylic acid, 5-bromo-, 1,1-dimethylethyl ester, tert-butyl5-bromo-1h-indazole-1-carboxylate, 1-tert-butoxycarbonyl-5-bromoindazole, 5-bromoindazole-1-carboxylic acid tert-butyl ester |
Formula Weight | 297.15 |
Percent Purity | 90% |
Chemical Name or Material | Tert-butyl 5-bromo-1H-indazole-1-carboxylate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'