missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl 4-[(trifluoromethyl)sulfonyloxy]-5,6-dihydropyridine-1(2h)-carboxylate, 90%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Spécification
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 331.31 |
Percent Purity | 97% |
Chemical Name or Material | Tert-butyl 4-[(trifluoromethyl)sulfonyloxy]-5, 6-dihydropyridine-1(2h)-carboxylate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'