missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-(aminocarbonyl)tetrahydropyridine-1(2H)-carboxylate, 97%, Maybridge

Brand:  Maybridge TL00951EA

Code : 31

Additional Details : CAS Number : 91419-48-6 Weight : 0.06000kg

Quantity 10g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10691123

  • £79.00 / 10g


Estimated Shipment date
from Supplier 27-03-2018
Add to basket

Specifications

Chemical Identifiers

Tert-butyl 4-(aminocarbonyl)tetrahydropyridine-1(2H)-carboxylate
97.0
97%
228.29
91419-48-6
97%
C11H20N2O3
MFCD02180953

Specifications

Tert-butyl 4-(aminocarbonyl)tetrahydropyridine-1(2H)-carboxylate
97.0
97%
C11H20N2O3
MFCD02180953
91419-48-6
97%
Amber Glass Bottle
228.29
10g
Videos
SDS