missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-{4-[(methylamino)methyl]pyrid-2-yl}piperazine-1-carboxylate, 97%, Thermo Scientific™

Catalog Number p-3586900
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
946409-15-0
C16H26N4O2
306.41
MFCD09966148
BNLNDKXJNWJYKP-UHFFFAOYSA-N
tert-butyl 4-4-methylamino methyl pyridin-2-yl piperazine-1-carboxylate, tert-butyl 4-4-methylamino methyl pyrid-2-yl piperazine-1-carboxylate
28765107
tert-butyl 4-[4-(methylaminomethyl)pyridin-2-yl]piperazine-1-carboxylate
CNCC1=CC(=NC=C1)N1CCN(CC1)C(=O)OC(C)(C)C
This item is not returnable. View return policy
946409-15-0
C16H26N4O2
306.41
MFCD09966148
BNLNDKXJNWJYKP-UHFFFAOYSA-N
tert-butyl 4-4-methylamino methyl pyridin-2-yl piperazine-1-carboxylate, tert-butyl 4-4-methylamino methyl pyrid-2-yl piperazine-1-carboxylate
28765107
tert-butyl 4-[4-(methylaminomethyl)pyridin-2-yl]piperazine-1-carboxylate
CNCC1=CC(=NC=C1)N1CCN(CC1)C(=O)OC(C)(C)C
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 306.41
Percent Purity 97%
Chemical Name or Material tert-Butyl 4-{4-[(methylamino)methyl]pyrid-2-yl}piperazine-1-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'